Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MMP-2, Mouse, Unlabeled, Clone: 75-7F7,

Catalog No. IM51100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
IM51100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IM51100UG Supplier MilliporeSigma™ Supplier No. IM51100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recognizes the ∽72kDa latent and the ∽66kDa active forms of MMP-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 75-7F7
Concentration Please refer to vial label for lot-specific concentration
Conjugate Unconjugated
Formulation In 100mM sodium phosphate buffer, 0.1% BSA, pH 7.0, ≤0.1% Sodium Azide.
Gene Alias Anti-72 kDa Gelatinase, Anti-Gelatinase A, Anti-Matrix Metalloproteinase 2
Host Species Mouse
Immunogen human MMP-2 derived from conditioned medium of CCD-41SK skin fibroblasts
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine
Content And Storage -20°C
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.