Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-20 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS0985R

Description
MMP20 degrades amelogenin, the major protein component of the enamel matrix, as well as two of the macromolecules characterizing the cartilage extracellular matrix: aggrecan and the cartilage oligomeric matrix protein (COMP). MMP20 may play a central role in tooth enamel formation.Specifications
MMP-20 | |
Polyclonal | |
Unconjugated | |
MMP20 | |
AI2A2; BOS_14256; Enamel metalloproteinase; Enamelysin; LOC100355428; matrix metallo protease; matrix metallopeptidase 20; matrix metallopeptidase 20 (enamelysin); matrix metallopeptidase 25; matrix metalloproteinase; matrix metalloproteinase 20; matrix metalloproteinase 20 (enamelysin); Matrix metalloproteinase-20; MMP; MMP20; MMP-20; MMP25; MMPs | |
Rabbit | |
Protein A | |
RUO | |
9313 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O60882 | |
MMP20 | |
KLH conjugated synthetic peptide derived from human MMP20 . | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction