Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-24/MT5-MMP Antibody (RM0289-7J71), Alexa Fluor™ 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211803AF405
Description
MMP-24/MT5-MMP Monoclonal specifically detects MMP-24/MT5-MMP in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
MMP-24/MT5-MMP | |
Monoclonal | |
Alexa Fluor 405 | |
EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 24 (membrane-inserted), matrix metalloproteinase 24 (membrane-inserted), matrix metalloproteinase-24, membrane-type 5 matrix metalloproteinase, Membrane-type matrix metalloproteinase 5, Membrane-type-5 matrix metalloproteinase, MMP-24, MT5MMP, MT5-MMPMMP25, MT-MMP 5, MTMMP5, MT-MMP5 | |
Rat | |
Protein A or G purified | |
RUO | |
10893 | |
Mouse | |
Purified |
Western Blot, Flow Cytometry | |
RM0289-7J71 | |
Western Blot, Flow Cytometry | |
MMP24 | |
Mouse recombinant MMP-24/MT5-MMP (extracellular domain) | |
0.1 mL | |
Primary | |
This antibody weakly cross reacts with mouse MMP3 and MMP-9. | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction