Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-24/MT5-MMP Antibody (RM0289-7J71), Alexa Fluor™ 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211803AF594
Description
MMP-24/MT5-MMP Monoclonal specifically detects MMP-24/MT5-MMP in Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
MMP-24/MT5-MMP | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
MMP24 | |
Mouse recombinant MMP-24/MT5-MMP (extracellular domain) | |
0.1 mL | |
10893.0 | |
Mouse | |
IgG2 |
Western Blot, Flow Cytometry | |
RM0289-7J71 | |
Western Blot, Flow Cytometry | |
EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 24 (membrane-inserted), matrix metalloproteinase 24 (membrane-inserted), matrix metalloproteinase-24, membrane-type 5 matrix metalloproteinase, Membrane-type matrix metalloproteinase 5, Membrane-type-5 matrix metalloproteinase, MMP-24, MT5MMP, MT5-MMPMMP25, MT-MMP 5, MTMMP5, MT-MMP5 | |
Rat | |
Protein A or G purified | |
Primary | |
This antibody weakly cross reacts with mouse MMP3 and MMP-9. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction