Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-25/MT6-MMP Antibody (141808), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1142J
This item is not returnable.
View return policy
Description
MMP-25/MT6-MMP Monoclonal antibody specifically detects MMP-25/MT6-MMP in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
MMP-25/MT6-MMP | |
Monoclonal | |
Janelia Fluor 646 | |
leukolysin, matrix metallopeptidase 25, matrix metallopeptidase-like 1, matrix metalloproteinase 20, matrix metalloproteinase 25, matrix metalloproteinase-25, matrix metalloproteinase-like 1, membrane-type 6 matrix metalloproteinase, membrane-type matrix metalloproteinase 6, membrane-type-6 matrix metalloproteinase, MMP20, MMP20A, MMP-25, MMPL1, MT6MMP, MT6-MMP, MT-MMP 6, MTMMP6, MT-MMP6 | |
Mouse myeloma cell line NS0-derived recombinant human MMP-25/MT6-MMP, Pro314-Cys508, Accession # Q9NPA2 | |
0.5 mL | |
Angiogenesis, Cancer | |
64386 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry | |
141808 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction