Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-25/MT6-MMP Antibody (MM0029-2B5), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11060995B
Description
MMP-25/MT6-MMP Monoclonal specifically detects MMP-25/MT6-MMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MMP-25/MT6-MMP | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
MMP25 | |
Human recombinant MMP-25/MT6-MMP | |
0.1 mL | |
Primary | |
MMP25 (MM0029-2B5) | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0029-2B5 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
leukolysin, matrix metallopeptidase 25, matrix metallopeptidase-like 1, matrix metalloproteinase 20, matrix metalloproteinase 25, matrix metalloproteinase-25, matrix metalloproteinase-like 1, membrane-type 6 matrix metalloproteinase, membrane-type matrix metalloproteinase 6, membrane-type-6 matrix metalloproteinase, MMP20, MMP20A, MMP-25, MMPL1, MT6MMP, MT6-MMP, MT-MMP 6, MTMMP6, MT-MMP6 | |
Mouse | |
Protein G purified | |
Angiogenesis, Cancer | |
64386 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction