Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-3 Antibody (50647), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB513N
Description
MMP-3 Monoclonal antibody specifically detects MMP-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
MMP-3 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF | |
50647 | |
Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1 | |
Chinese hamster ovary cell line CHO-derived recombinant human MMP-3, Tyr18-Cys477, Accession # P08254 | |
0.5 mL | |
Angiogenesis, Cancer, Extracellular Matrix | |
4314 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction