Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-3 Antibody (MMP3/2655), Alexa Fluor™ 488, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP279865AF488

Catalog No. NB011363


Only null left
Add to Cart

Description

Description

MMP-3 Monoclonal specifically detects MMP-3 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

MMP-3
Monoclonal
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
MMP3
Recombinant fragment of human MMP-3 protein (around aa 236-363) (exact sequence is proprietary) (Uniprot: P08254)
0.1 mL
Angiogenesis, Cancer, Extracellular Matrix
Human
IgG2b κ
Western Blot, ELISA
MMP3/2655
Western Blot, ELISA
CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1
Mouse
Protein A purified
RUO
Primary
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.