Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-3 Antibody (MMP3/2655), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279865FR
Description
MMP-3 Monoclonal specifically detects MMP-3 in Human samples. It is validated for Western Blot, ELISA.Specifications
MMP-3 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MMP3 | |
Recombinant fragment of human MMP-3 protein (around aa 236-363) (exact sequence is proprietary) (Uniprot: P08254) | |
0.1 mL | |
Angiogenesis, Cancer, Extracellular Matrix | |
Human | |
IgG2b κ |
Western Blot, ELISA | |
MMP3/2655 | |
Western Blot, ELISA | |
CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction