Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-3 Antibody (MMP3/2806), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308626G
Description
MMP-3 Monoclonal antibody specifically detects MMP-3 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein ArraySpecifications
MMP-3 | |
Monoclonal | |
DyLight 488 | |
CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1 | |
Recombinant full-length human MMP-3 protein (Uniprot: P08254 ) | |
100 μg | |
Angiogenesis, Cancer, Extracellular Matrix | |
4314 | |
Store at 4°C in the dark. | |
IgG2c κ |
ELISA, Immunohistochemistry (Paraffin), Protein Array | |
MMP3/2806 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction