Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-3 Rabbit anti-Human, Clone: MMP3/1994R, Novus Biologicals™

Rabbit Monoclonal Antibody
$304.50
Specifications
Antigen | MMP-3 |
---|---|
Clone | MMP3/1994R |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA |
Applications | Western Blot, Flow Cytometry, ELISA |
Description
MMP-3 Monoclonal specifically detects MMP-3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
MMP-3 | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Extracellular Matrix | |
10 mM PBS with 0.05% BSA | |
4314 | |
IgG | |
Protein A purified |
MMP3/1994R | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1 | |
Recombinant fragment (around aa 317-327) of human MMP-3 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title