Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-7 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP199123AF350
Description
MMP-7 Polyclonal specifically detects MMP-7 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MMP-7 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 3.4.24, EC 3.4.24.23, matrilysin, matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
Rabbit | |
Affinity Purified | |
Angiogenesis, Cancer, Cell Biology, Cytoskeleton Markers, Extracellular Matrix, Wnt Signaling Pathway | |
4316.0 | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
MMP7 | |
A synthetic peptide made to an internal portion of the human MMP7 protein (between residues 250-300) [UniProt P09237] | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction