Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NLRP3/NALP3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP212446
Description
NLRP3/NALP3 Polyclonal specifically detects NLRP3/NALP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunomicroscopy, Knockdown Validated.Specifications
NLRP3/NALP3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, anti-mouse NLRP3, anti-NLRP3, AVP, C1orf7, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, NLRP3 mouse, NLRP3 polyclonal, NLRP3 rat, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1 | |
Rabbit | |
118 kDa | |
0.2 mL | |
Alzheimers Research, Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Inflammation, Neurodegeneration, Signal Transduction | |
114548 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunomicroscopy, KnockDown | |
1.0 mg/mL | |
Western Blot 2 - 5 ug/mL, Flow Cytometry reported in scientific literature (PMID 34993560), Immunohistochemistry 1:10 - 1:50, Immunocytochemistry/Immunofluorescence, Immunoprecipitation reported in scientific literature (PMID 37029500), Immunohistochemistry-Paraffin 1:10 - 1:50, Immunohistochemistry-Frozen reported in scientific literature (PMID 35792172), Immunomicroscopy, KnockDown Validated | |
Q96P20 | |
NLRP3 | |
This NLRP3/NALP3 antibody was raised against a portion of amino acids 1-50 of human NLRP3/NALP3. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Reacts with Human, Mouse, New World Monkey and Rat. The amino acid sequence used as immunogen is 100% homologous in Human and Monkey and 86% homologous in Mouse and Rat. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction