Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NLRP3/NALP3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-5085964 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212446SS 0.05 mL
NBP212446 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP212446SS Supplier Novus Biologicals Supplier No. NBP212446SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NLRP3/NALP3 Polyclonal specifically detects NLRP3/NALP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunomicroscopy, Knockdown Validated.

Specifications

Antigen NLRP3/NALP3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunomicroscopy, KnockDown
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 - 5 ug/mL, Flow Cytometry reported in scientific literature (PMID 34993560), Immunohistochemistry 1:10 - 1:50, Immunocytochemistry/Immunofluorescence, Immunoprecipitation reported in scientific literature (PMID 37029500), Immunohistochemistry-Paraffin 1:10 - 1:50, Immunohistochemistry-Frozen reported in scientific literature (PMID 35792172), Immunomicroscopy, KnockDown Validated
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q96P20
Gene Alias AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, anti-mouse NLRP3, anti-NLRP3, AVP, C1orf7, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, NLRP3 mouse, NLRP3 polyclonal, NLRP3 rat, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1
Gene Symbols NLRP3
Host Species Rabbit
Immunogen This NLRP3/NALP3 antibody was raised against a portion of amino acids 1-50 of human NLRP3/NALP3.
Molecular Weight of Antigen 118 kDa
Purification Method Immunogen affinity purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Inflammation, Neurodegeneration, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 114548
Test Specificity Reacts with Human, Mouse, New World Monkey and Rat. The amino acid sequence used as immunogen is 100% homologous in Human and Monkey and 86% homologous in Mouse and Rat.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.