Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-7 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP199123F
Description
MMP-7 Polyclonal specifically detects MMP-7 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MMP-7 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
P09237 | |
MMP7 | |
A synthetic peptide made to an internal portion of the human MMP7 protein (between residues 250-300) [UniProt P09237] | |
0.1 mL | |
Angiogenesis, Cancer, Cell Biology, Cytoskeleton Markers, Extracellular Matrix, Wnt Signaling Pathway | |
4316 | |
Store at 4C in the dark. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS with 0.05% Sodium Azide | |
EC 3.4.24, EC 3.4.24.23, matrilysin, matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction