Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-7 Antibody (MM0022-4C21), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11060988MFV450
Description
MMP-7 Monoclonal antibody specifically detects MMP-7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MMP-7 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0022-4C21 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 3.4.24, EC 3.4.24.23, matrilysin, matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
Human recombinant MMP-7 | |
0.1 mL | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Wnt Signaling Pathway | |
4316 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction