Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-8 Antibody (MM0023-7A11), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11060989B
Description
MMP-8 Monoclonal specifically detects MMP-8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MMP-8 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
MMP8 | |
Human recombinant MMP-8 | |
0.1 mL | |
Primary | |
This antibody recognizes human MMP-8 in western blot without cross reactivity with other MMPs. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0023-7A11 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CLG1HNC, EC 3.4.24, EC 3.4.24.34, matrix metallopeptidase 8 (neutrophil collagenase), matrix metalloproteinase 8 (neutrophil collagenase), Matrix metalloproteinase-8, MMP-8, neutrophil collagenase, PMNL collagenase, PMNL-CL | |
Mouse | |
Protein G purified | |
Cancer | |
4317 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction