Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-9 (92kDa Collagenase IV) Ab-4 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS816P0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS816P0 100 μL 200μg/mL
MS816P1ABX 100 μL 1mg/mL
MS816P1 500 μL 200μg/mL
MS816P 1 mL 200μg/mL
MS816PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS816P0 Supplier Epredia Supplier No. MS816P0

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Epredia™ MMP-9 (92kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

MMPs are a group of enzymes involved in matrix degradation. They share some important characteristics, such as a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and inhibition by specific proteinase inhibitors known as tissue inhibitors of metalloproteinases (TIMPs). Reportedly, MMPs play a crucial role in tumor cell invasion and metastasis.

Host Species: Mouse

Clone: IA5

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human native 92kDa MMP-9

Molecular Weight: 92kDa (proform) and ∽86kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human fibrosarcoma HT-1080 cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Not suitable)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen MMP-9 (92kDa Collagenase IV) Ab-4
Applications Immunofluorescence
Classification Monoclonal
Clone IA5
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human native 92kDa MMP-9
Quantity 100 μL
Regulatory Status RUO
Research Discipline Proteases And Inhibitors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.