Learn More
Description
Host Species: Mouse
Clone: IA5
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Human native 92kDa MMP-9
Molecular Weight: 92kDa (proform) and ∽86kDa (active form)
Positive Control: Conditioned, serum-free medium from (TPA-treated) human fibrosarcoma HT-1080 cells
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Not suitable)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Not suitable)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | MMP-9 (92kDa Collagenase IV) Ab-4 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | IA5 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human native 92kDa MMP-9 |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.