Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MMP-9, Active, Human, Recombinant Protein

Catalog No. pf0245ug Shop All MilliporeSigma Products
Click to view available options
Quantity:
5 μg

Catalog No. PF0245UG

Supplier: MilliporeSigma™ PF0245UG

Only null left
Add to Cart
Add to Cart
Recombinant, human MMP-9 purified from transfected mammalian cells and activated with APMA. Active MMP-9 enzyme is APMA-free.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 50mM HEPES, 10mM CaCl2, 20% glycerol, 0.005% BRIJ™-35 Detergent, pH 7.5.
Purification Method SDS-PAGE
Quantity 5 μg
Immunogen MMP-9
Storage Requirements Following initial use, aliquot into siliconized vials and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.