Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MMP-9, Active, Human, Recombinant Protein

Catalog No. pf1405ug
Click to view available options
Quantity:
5 μg
Recombinant, human MMP-9 with a truncated C-terminal hemopexin domain expressed as proenzyme that is activated by APMA. AMPA is removed through a Biogel-P6 column and active MMP-9 is purified.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
For Use With (Application) Immunoblotting, Zymography
Formulation In 50mM Hepes, 10mM CaCl2, 20% glycerol, 0.005% BRIJ™ 35 Detergent, pH 7.5.
Quantity 5 μg
Immunogen MMP-9, Active
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.