Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-9 Antibody (043), Alexa Fluor™ 405

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289415AF405
Description
MMP-9 Monoclonal specifically detects MMP-9 in Human samples. It is validated for Sandwich ELISA Detection.Specifications
| MMP-9 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 43 | |
| Sandwich ELISA Detection | |
| 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human MMP-9 (NP_004985.2; Met1-Asp707). | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR | |
| 4318 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction