Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-9 Antibody (213), DyLight 594

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290868DL594
Description
MMP-9 Monoclonal specifically detects MMP-9 in Rat samples. It is validated for ELISA.Specifications
MMP-9 | |
Monoclonal | |
DyLight 594 | |
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat MMP-9 (EDL96479.1; Met1-Pro708). | |
0.1 mL | |
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR | |
4318 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
213 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction