Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-9 Antibody (4A3), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 16 publications

$206.50 - $501.50

Specifications

Antigen MMP-9
Clone 4A3
Concentration 1.0 mg/mL
Dilution Western Blot 1:500 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP213173SS
SDP
View Documents
Novus Biologicals
NBP213173SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP213173
SDP
View Documents
Novus Biologicals
NBP213173
0.1 mL
Each for $501.50
Only null left
Add to Cart
 
Description

Description

MMP-9 Monoclonal specifically detects MMP-9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

MMP-9
1.0 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR
P14780
4318
Synthetic peptide from the N-terminus of human MMP9 [Swiss-Prot# P14780]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
4A3
Western Blot 1:500 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
MMP9
IgG1 κ
Protein G purified
Active MMP9. It does not react with the MMP9 proenzyme or the active and proenzyme forms of MMP2.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.