Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-9 Antibody (S51-82), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$560.50
Specifications
Antigen | MMP-9 |
---|---|
Clone | S51-82 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Description
MMP-9 Monoclonal specifically detects MMP-9 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
MMP-9 | |
1 mg/mL | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR | |
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
MMP9 | |
IgG2a | |
Protein G purified |
S51-82 | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
4318 | |
Fusion protein amino acids 1-708 (full length) of rat MMP9 | |
Primary | |
Detecs ∼92kDa and ∼82kDa (pro and active forms). |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title