Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-9 Antibody (V6P3F11*A4), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250515C
Description
MMP-9 Monoclonal specifically detects MMP-9 in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
MMP-9 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MMP9 | |
Ovalbumin-conjugated synthetic peptide KAQLIDRGYP (C-terminal sequence) | |
0.1 mL | |
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR | |
4318 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
V6P3F11*A4 | |
Western Blot, ELISA, Immunohistochemistry | |
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction