Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-9 Mouse anti-Human, Mouse, HRP, Clone: SB15c, Southern Biotech™
SDP

Catalog No. OB1202505 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
1.0 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB1202505 1.0 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB1202505 Supplier Southern Biotechnology Associates Supplier No. 1202505

Mouse Monoclonal Antibody

SouthernBiotech is a leading provider of antibodies and proteins for extracellular matrix research.

Specifications

Antigen MMP-9
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone SB15c
Concentration Lot specific
Conjugate HRP
Formulation Supplied in a stock solution in 50% glycerol/50% PBS, pH 7.4; No preservative added
Host Species Mouse
Immunogen Recombinant full length MMP-9
Quantity 1.0 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 2-8°C
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.