Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-9 Rabbit anti-Human, Clone: 43, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | MMP-9 |
---|---|
Clone | 43 |
Dilution | Sandwich ELISA Detection 1:1000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
MMP-9 Monoclonal specifically detects MMP-9 in Human samples. It is validated for Sandwich ELISA Detection.Specifications
MMP-9 | |
Sandwich ELISA Detection 1:1000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase | |
Recombinant Human MMP-9 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
43 | |
ELISA | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR | |
0.2 um filtered solution in PBS | |
4318 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title