Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-9, Rabbit anti-Rat, Human, Polyclonal, Millipore Sigma™

Catalog No. ABT54425UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT54425UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT54425UL Supplier MilliporeSigma Supplier No. ABT54425UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-MMP-9, Cat. No. ABT544, is a rabbit polyclonal antibody that detects Matrix Metalloroteinase-9 and is tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Matrix metalloproteinase-9 (UniProt: P50282; also known as EC: 3.4.24.35, MMP-9, 92 kDa gelatinase, 92 kDa type IV collagenase, Gelatinase B, GELB) is encoded by the Mmp9 gene (Gene ID: 81687) in rat. Matrix Metalloproteinases (MMPs) are a family of zinc metallo-endopeptidases secreted by cells and are responsible for much of the turnover of matrix components. MMP-9 is a secreted extracellular matrix metalloproteinase that exhibits a broad range of substrate specificity for native collagens including types IV, V, VII, and X as well as gelatin, proteoglycans, and elastin. It can exist as monomer or disulfide-linked homodimer. MMP-9 is synthesized with a signal peptide (aa 1-19) and a propeptide (aa 20-107) that are subsequently cleaved to produce active enzyme. Its activity can be inhibited by TIMP-1, which binds exclusively to pro-MMP-9 (Kd ∽35 nM). MMP-9 contain a fibronectin-like domain inserted into the catalytic domain, presumably to enhance substrate binding. MMP-9 also contains a collagen type-V-like domain, which may enhance substrate binding and specificity. MMP-9 contains a cysteine switch (aa 98-105) where the conserved cysteine binds the catalytic zinc ion and inhibits its enzyme activity. The dissociation of the cysteine from the zinc ion upon the activation-peptide release activates the enzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP-9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected MMP-9 in rat spleen and human bone marrow tissues.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Matrix metalloproteinase-9;EC: 3.4.24.35;92 kDa gelatinase;92 kDa type IV collagenase;Gelatinase B;GELB
Gene Symbols Mmp9
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal half of rat Matrix metalloproteinase-9.
Molecular Weight of Antigen ∽85 kDa obsereved; 78.61 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects human and rat Matrix metalloproteinase-9. It targets an epitope within 16 amino acids from the N-terminal half.
Target Species Rat, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.