Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP10 Monoclonal Antibody (IVC5), Invitrogen™

Catalog No. PIMA514233
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514233 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514233 Supplier Thermo Scientific Supplier No. MA514233

Mouse Monoclonal Antibody

MA5-14233 targets MMP-10 (Stromelysin-2) in IP and WB applications and shows reactivity with Human samples. The MA5-14233 immunogen is human recombinant MMP-10 (stromelysin-2 protein).

MMP10 belongs to the matrix metalloproteinase family, and is generally secreted in its pro form, which is further modified by extracellular proteinases. As a metalloproteinase, MMP10 requires zinc and calcium as cofactors for activity. It is involved in the activation of pro-collagenase, catabolism of collagen, degradation of fibronectin, and tissue remodeling. In humans, the gene MMP10 located on chromosome 11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP10
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone IVC5
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene MMP10
Gene Accession No. P09238
Gene Alias AV377895; matrix metallo protease; matrix metallopeptidase 10; matrix metallopeptidase 10 (stromelysin 2); matrix metalloprotease 10; matrix metalloproteinase 10; matrix metalloproteinase 10 (stromelysin 2); matrix metalloproteinase-10; MMP; Mmp10; MMP-10; MMPs; SL-2; STMY2; stromelysin 2; stromelysin-2; Transformation-associated protein 34A; transin 2; transin-2
Gene Symbols MMP10
Host Species Mouse
Immunogen Human recombinant MMP-10 (stromelysin-2 protein)
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4319
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.