Learn More
Invitrogen™ MMP10 Polyclonal Antibody

Description
Positive test controls include: BT-474, MCF7, U-251MG, M21. The target is usually found in the following locations: Secreted, extracellular matrix, extracellular space. Immunogen sequence: PLYNSFTELA QFRLSQDDVN GIQSLYGPPP ASTEEPLVPT KSVPSGSEMP AKCDPALSFD AISTLRGEYL FFKDRYFWRR SHWNPEPEFH LISAFWPSLP SYLDAAYEVN SRDTVFIFKG NEFWAIRGNE VQAGYPRGIH TLGFPPTIRK IDAAVSDKEK KKTYFFAADK YWRFDENSQS MEQGFPRLIA DDFPGVEPKV DAVLQAFGFF YFFSGSSQFE FDPNARMVTH ILKSNSWLHC.
Specifications
Specifications
| Antigen | MMP10 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2.01 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MMP10 |
| Gene Accession No. | O55123, P07152, P09238 |
| Gene Alias | AV377895; matrix metallo protease; matrix metallopeptidase 10; matrix metallopeptidase 10 (stromelysin 2); matrix metalloprotease 10; matrix metalloproteinase 10; matrix metalloproteinase 10 (stromelysin 2); matrix metalloproteinase-10; MMP; Mmp10; MMP-10; MMPs; SL-2; STMY2; stromelysin 2; stromelysin-2; Transformation-associated protein 34A; transin 2; transin-2 |
| Gene Symbols | MMP10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.