Learn More
Invitrogen™ MMP11 Polyclonal Antibody

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat spleen tissue, MM231 whole cell. IHC: mouse spleen tissue, rat spleen tissue, human appendicitis tissue.
Specifications
Specifications
| Antigen | MMP11 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and 0.05mg sodium azide |
| Gene | Mmp11 |
| Gene Accession No. | P24347, Q02853, Q499S5 |
| Gene Alias | matrix metallo protease; matrix metallopeptidase 11; matrix metallopeptidase 11 (stromelysin 3); Matrix metalloproteinase 11 (stromelysin 3); matrix metalloproteinase-11; MMP; MMP11; MMP-11; MMPs; SL-3; ST3; Stmy3; stromelysin III; stromelysin-3 |
| Gene Symbols | Mmp11 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.