Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMP13 Monoclonal Antibody (C2)

Catalog No. PIMA543722
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543722 20 μL
PIMA541638 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543722 Supplier Invitrogen™ Supplier No. MA543722
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMP13 Monoclonal Antibody for Western Blot

MMP13 (collagense 3) plays a role in the degradation of extracellular matrix proteins including fibrillar collagen, fibronectin, TNC, and ACAN. MMP13 cleaves tripal helical collagens, including type I, II, and III collagen - but has the highest activity with soluble type II collagen. It can degrade collagen type IV, XIV, and X. MMP13 plays a role in wound healing, tissue remodeling, cartilage degradation, bone development, bone mineralization, and ossification. It is required for normal embryonic bone development and ossificaiton. Mutations affecting the gene can lead to spondyloepimetaphyseal dysplasia Missouri type, metaphyseal anadysplasia 1, and metaphyseal dysplasia Sphar type.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP13
Applications Western Blot
Classification Monoclonal
Clone C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene MMP13
Gene Accession No. P45452
Gene Alias cb1034; Clg; CLG3; Collagenase; collagenase 3; collagenase 3a; matrix metalloproteinase 13a; collagenase-1; Collagenase3; Collagenase-3; collagenase-3 precursor; interstitial collagenase; MANDP1; matrix metallo protease; matrix metallopeptidase 13; matrix metallopeptidase 13 (collagenase 3); matrix metallopeptidase 13a; matrix metallopeptidase 13a (collagenase 3); matrix metalloproteinase 13; matrix metalloproteinase 13 (collagenase 3); matrix metalloproteinase-13; matrix metalloproteinase-13; MMP-13; interstitial protein; MMP; Mmp1; MMP13; MMP-13; mmp13 protein; mmp13a; MMPLg; MMPLh; MMPs; Procollagenase 3; UMRCASE; ZFMMP13
Gene Symbols MMP13
Host Species Mouse
Immunogen Recombinant protein Matrix Metalloproteinase 13. The antigen corresponds to amino acid range 103-290 of the target protein.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4322
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.