Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MMP13 Polyclonal Antibody
Guinea Pig Polyclonal Antibody
Supplier: Invitrogen™ PA5121879
This item is not returnable.
View return policy
Description
MMP13 Polyclonal Antibody for Western Blot
MMP13 (collagense 3) plays a role in the degradation of extracellular matrix proteins including fibrillar collagen, fibronectin, TNC, and ACAN. MMP13 cleaves tripal helical collagens, including type I, II, and III collagen - but has the highest activity with soluble type II collagen. It can degrade collagen type IV, XIV, and X. MMP13 plays a role in wound healing, tissue remodeling, cartilage degradation, bone development, bone mineralization, and ossification. It is required for normal embryonic bone development and ossificaiton. Mutations affecting the gene can lead to spondyloepimetaphyseal dysplasia Missouri type, metaphyseal anadysplasia 1, and metaphyseal dysplasia Sphar type.
Specifications
MMP13 | |
Polyclonal | |
Unconjugated | |
MMP13 | |
cb1034; Clg; CLG3; Collagenase; collagenase 3; collagenase 3a; matrix metalloproteinase 13a; collagenase-1; Collagenase3; Collagenase-3; collagenase-3 precursor; interstitial collagenase; MANDP1; matrix metallo protease; matrix metallopeptidase 13; matrix metallopeptidase 13 (collagenase 3); matrix metallopeptidase 13a; matrix metallopeptidase 13a (collagenase 3); matrix metalloproteinase 13; matrix metalloproteinase 13 (collagenase 3); matrix metalloproteinase-13; matrix metalloproteinase-13; MMP-13; interstitial protein; MMP; Mmp1; MMP13; MMP-13; mmp13 protein; mmp13a; MMPLg; MMPLh; MMPs; Procollagenase 3; UMRCASE; ZFMMP13 | |
Guinea Pig | |
Antigen affinity chromatography | |
RUO | |
100008685 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
O62806 | |
MMP13 | |
Recombinant protein Matrix Metalloproteinase 13. The antigen corresponds to amino acid range 70-245 of the target protein. | |
20 μL | |
Primary | |
Rabbit | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction