Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP13 Polyclonal Antibody, HRP, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS10581RHRP

This item is not returnable.
View return policy
Description
MMP13 Polyclonal antibody specifically detects MMP13 in Human, Mouse, Porcine, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western BlotSpecifications
MMP13 | |
Polyclonal | |
HRP | |
MMP13 | |
cb1034; Clg; CLG3; Collagenase; collagenase 3; collagenase 3a; matrix metalloproteinase 13a; collagenase-1; Collagenase3; Collagenase-3; collagenase-3 precursor; interstitial collagenase; MANDP1; matrix metallo protease; matrix metallopeptidase 13; matrix metallopeptidase 13 (collagenase 3); matrix metallopeptidase 13a; matrix metallopeptidase 13a (collagenase 3); matrix metalloproteinase 13; matrix metalloproteinase 13 (collagenase 3); matrix metalloproteinase-13; matrix metalloproteinase-13; MMP-13; interstitial protein; MMP; Mmp1; MMP13; MMP-13; mmp13 protein; mmp13a; MMPLg; MMPLh; MMPs; Procollagenase 3; UMRCASE; ZFMMP13 | |
KLH conjugated synthetic peptide derived from human MMP13 | |
100 μL | |
Primary | |
Human, Mouse, Pig, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
P23097, P33435, P45452 | |
Rabbit | |
Protein A | |
RUO | |
171052, 17386, 397346, 4322 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction