Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP13 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS10250R

Description
MMP13 (collagense 3) plays a role in the degradation of extracellular matrix proteins including fibrillar collagen, fibronectin, TNC, and ACAN. MMP13 cleaves tripal helical collagens, including type I, II, and III collagen - but has the highest activity with soluble type II collagen. It can degrade collagen type IV, XIV, and X. MMP13 plays a role in wound healing, tissue remodeling, cartilage degradation, bone development, bone mineralization, and ossification. It is required for normal embryonic bone development and ossificaiton. Mutations affecting the gene can lead to spondyloepimetaphyseal dysplasia Missouri type, metaphyseal anadysplasia 1, and metaphyseal dysplasia Sphar type.Specifications
MMP13 | |
Polyclonal | |
Unconjugated | |
MMP13 | |
cb1034; Clg; CLG3; Collagenase; Collagenase 3; collagenase-1; Collagenase3; collagenase-3; interstitial collagenase; MANDP1; matrix metallo protease; matrix metallopeptidase 13; matrix metallopeptidase 13 (collagenase 3); matrix metallopeptidase 13a; matrix metallopeptidase 13a (collagenase 3); matrix metalloproteinase 13; matrix metalloproteinase 13 (collagenase 3); matrix metalloproteinase 13a; Matrix metalloproteinase-13; MMP; Mmp1; Mmp13; MMP-13; mmp13 protein; mmp13a; MMPLg; MMPLh; MMPs; Procollagenase 3; UMRCASE; ZFMMP13 | |
Rabbit | |
Protein A | |
RUO | |
4322 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P45452 | |
MMP13 | |
KLH conjugated synthetic peptide derived from human MMP13. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction