Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP14 Mouse anti-Human, Clone: 3A6, Bioss
SDP

Catalog No. 501979804
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9804 Supplier Bioss Supplier No. BSM51028M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP′s are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. However, the protein encoded by this gene is a member of the membrane-type MMP (MT-MMP) subfamily; each member of this subfamily contains a potential transmembrane domain suggesting that these proteins are expressed at the cell surface rather than secreted. This protein activates MMP2 protein, and this activity may be involved in tumor invasion.

Specifications

Antigen MMP14
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3A6
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene MMP14
Gene Accession No. P50281
Gene Alias AI325305; cb386; EC 3.4.24.80; matrix metallo protease; matrix metallopeptidase 14; matrix metallopeptidase 14 (membrane-inserted); matrix metallopeptidase 14a (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted) alpha; matrix metalloproteinase 14 membrane-inserted; matrix metalloproteinase 14, membrane-inserted; matrix metalloproteinase 14a (membrane-inserted); Matrix metalloproteinase-14; membrane type 1 metalloprotease; membrane type 1-matrix metalloproteinase; Membrane type 1-MMP; Membrane-type matrix metalloproteinase 1; membrane-type matrix metalloproteinase 1 alpha; Membrane-type-1 matrix metalloproteinase; MMP; MMP14; MMP-14; mmp14a; MMPs; MMP-X MT-MMP; MMP-X1; MT1MMP; MT1-MMP; MT1-MMP-alpha; Mtmmp; MT-MMP; MT-MMP 1; MT-MMP MT1-MMP; MTMMP1; MT-MMP-1; MT-MMPalpha; sabe; sb:cb386; type 1 matrix metalloprotease 14; WNCHRS; wu:fb72b09; zMT-MMPalpha; zMT-MMP-alpha
Gene Symbols MMP14
Host Species Mouse
Immunogen This MMP14 antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 470-499 amino acids from the C-terminal region of human MMP14.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4323
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.