Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMP14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585741 Supplier Invitrogen™ Supplier No. PA585741
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Pig (100%), Sheep (100%), Rhesus Monkey (100%), Bovine (100%). Positive Control: U87-MG, Neuro2A, MMP14-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. However, the protein encoded by this gene is a member of the membrane-type MMP (MT-MMP) subfamily; each member of this subfamily contains a potential transmembrane domain suggesting that these proteins are expressed at the cell surface rather than secreted. This protein activates MMP2 protein, and this activity may be involved in tumor invasion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP14
Applications Western Blot
Classification Polyclonal
Concentration 1.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene MMP14
Gene Accession No. P50281, P53690, Q10739
Gene Alias AI325305; cb386; EC 3.4.24.80; matrix metallo protease; matrix metallopeptidase 14; matrix metallopeptidase 14 (membrane-inserted); matrix metallopeptidase 14a (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted); matrix metalloproteinase 14 (membrane-inserted) alpha; matrix metalloproteinase 14 membrane-inserted; matrix metalloproteinase 14, membrane-inserted; matrix metalloproteinase 14a (membrane-inserted); matrix metalloproteinase-14; matrix metalloproteinase-14a; membrane type 1 metalloprotease; membrane type 1-matrix metalloproteinase; Membrane type 1-MMP; membrane-type matrix metalloproteinase 1; membrane-type matrix metalloproteinase 1 alpha; membrane-type-1 matrix metalloproteinase; MMP; MMP14; MMP-14; mmp14a; MMPs; MMP-X MT-MMP; MMP-X1; MT1MMP; MT1-MMP; MT1-MMP-alpha; Mtmmp; MT-MMP; MT-MMP 1; MT-MMP MT1-MMP; MTMMP1; MT-MMP-1; MT-MMPalpha; sabe; sb:cb386; type 1 matrix metalloprotease 14; WNCHRS; wu:fb72b09; zMT-MMPalpha; zMT-MMP-alpha
Gene Symbols MMP14
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human MMP14. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17387, 4323, 81707
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.