Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMP16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115582
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115582 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115582 Supplier Invitrogen™ Supplier No. PA5115582
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MMP16.

Matrix metallopeptidase 16 during embryonic development, reproduction, and tissue remodeling. MMP-16 is also involved in the ECM degradation processes associated with arthritis and metastasis. In humans, MMP-16 is a part of a cluster of MMP genes at the 11q22.3 locus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP16
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Mmp16
Gene Accession No. O35548, P51512, Q9WTR0
Gene Alias C8orf57; DKFZp761D112; matrix metallo protease; matrix metallopeptidase 16; matrix metallopeptidase 16 (membrane-inserted); matrix metalloproteinase 16; matrix metalloproteinase-16; Membrane type 3-MMP; membrane-type matrix metalloproteinase 3; Membrane-type-3 matrix metalloproteinase; MMP; MMP-1; Mmp16; MMP-16; MMPs; MMPX2; MMP-X2; Mt3mmp; MT3-MMP; MT-MMP 3; MT-MMP2; MTMMP3; MT-MMP3; Putative transmembrane protein C8orf57
Gene Symbols Mmp16
Host Species Rabbit
Immunogen A synthesized peptide derived from human MMP16(Accession P51512), corresponding to amino acid residues K563-V607.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17389, 4325, 65205
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.