Learn More
Invitrogen™ MMP16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528966
Description
Recommended positive controls: HepG2, mouse kidney, rat kidney, Rat dorsal root ganglion. Predicted reactivity: Mouse (98%), Rat (98%), Japanese Medaka (83%), Xenopus laevis (92%), Chicken (97%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Matrix metallopeptidase 16 during embryonic development, reproduction, and tissue remodeling. MMP-16 is also involved in the ECM degradation processes associated with arthritis and metastasis. In humans, MMP-16 is a part of a cluster of MMP genes at the 11q22.3 locus.
Specifications
| MMP16 | |
| Polyclonal | |
| Unconjugated | |
| Mmp16 | |
| C8orf57; DKFZp761D112; matrix metallo protease; matrix metallopeptidase 16; matrix metallopeptidase 16 (membrane-inserted); matrix metalloproteinase 16; matrix metalloproteinase-16; Membrane type 3-MMP; membrane-type matrix metalloproteinase 3; Membrane-type-3 matrix metalloproteinase; MMP; MMP-1; Mmp16; MMP-16; MMPs; MMPX2; MMP-X2; Mt3mmp; MT3-MMP; MT-MMP 3; MT-MMP2; MTMMP3; MT-MMP3; Putative transmembrane protein C8orf57 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17389, 4325, 65205 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O35548, P51512, Q9WTR0 | |
| Mmp16 | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human MMP16. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.