Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP2 Monoclonal Antibody (2B10D1), CoraLite488, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc CL48866366100UL


This item is not returnable.
View return policy
Description
MMP2 Monoclonal antibody specifically detects MMP2 in Human, Mouse, Rat samples. It is validated for ImmunocytochemistrySpecifications
MMP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q6P7Q1, Q8K3W0, Q9NXR7 | |
Mouse | |
100 μL | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles, store in dark | |
IgG1 |
Immunocytochemistry | |
2B10D1 | |
CoraLite 488 | |
BABAM2 | |
72 kDa gelatinase; 72 kDa type IV collagenase; 72 kDa type IV collagenase isoform a preproprotein; 72kD gelatinase; 72kD type IV collagenase; 72kDa gelatinase; 72kDa type IV collagenase; CLG 4A; CLG4; CLG4A; Collagenase; collagenase type IV-A; endopeptidase; GelA; gelatinase A; Gelatinase alpha; Mat; matrix metallo protease; matrix metallopeptidase 2; matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase); matrix metalloprotease 2; matrix metalloproteinase; matrix metalloproteinase 2; matrix metalloproteinase 2 (72 KDa type IV collagenase); matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase); matrix metalloproteinase-2; matrix metalloproteinase-II; MMP; MMP2; MMP-2; MMP-2 protein; MMP-II; MMPs; MONA; neutrophil gelatinase; PEX; prepro-72kDa matrix metalloproteinase; Progelatinase A; TBE1; TBE-1; wu:fa99h12; wu:fk89d01 | |
Protein G | |
RUO | |
107976, 362704, 9577 | |
Liquid | |
Human|Mouse|Rat |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction