Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP2 Monoclonal Antibody (CA-4001 (CA719E3C)), Invitrogen™

Catalog No. PIMA513590
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA513590 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA513590 Supplier Thermo Scientific Supplier No. MA513590
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-13590 targets MMP-2 (72kDa Collagenase IV) in IA and WB applications and shows reactivity with Human, mouse, and Rat samples. The MA5-13590 immunogen is n-terminal peptide APSPIIKFPGD-VAPKTDK of procollagenase IV.

MMP (matrix metalloproteinase) are proteolytic enzymes capable of degrading connective tissue components. MMP have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPa and TIMPs play a significant role in regulating angiogenesis. MMP2 is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids, and contains the basic structure of propeptide, catalytic, and hemopexin domains. The matrix metalloproteinases (MMPs) are a family of at least eighteen secreted and membrane-bound zincendopeptidases. Collectively, these enzymes can degrade all the components of the extracellular matrix, including fibrillar and non-fibrillar collagens, fibronectin, laminin and basement membrane glycoproteins. In general, a signal peptide, a propeptide, and a catalytic domain containing the highly conserved zinc-binding site characterizes the structure of the MMPs. Functionally, MMP2 is involved in tissue remodeling. Mutations in MMP-2 gene have been associated with Winchester syndrome and Nodulosis-Arthropathy-Osteolysis (NAO) syndrome. Two transcript variants encoding different isoforms of MMP-2 have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP2
Applications Immunohistochemistry (Paraffin), Inhibition Assays, Western Blot
Classification Monoclonal
Clone CA-4001 (CA719E3C)
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene MMP2
Gene Accession No. P08253, P33434, P33436
Gene Alias 72 kDa gelatinase; 72 kDa type IV collagenase; 72 kDa type IV collagenase isoform a preproprotein; 72kD gelatinase; 72kD type IV collagenase; 72kDa gelatinase; 72kDa type IV collagenase; CLG 4A; CLG4; CLG4A; Collagenase; collagenase type IV-A; endopeptidase; GelA; gelatinase A; Gelatinase alpha; Mat; matrix metallo protease; matrix metallopeptidase 2; matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase); matrix metalloprotease 2; matrix metalloproteinase; matrix metalloproteinase 2; matrix metalloproteinase 2 (72 KDa type IV collagenase); matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase); matrix metalloproteinase-2; matrix metalloproteinase-II; MMP; MMP2; MMP-2; MMP-2 protein; MMP-II; MMPs; MONA; neutrophil gelatinase; PEX; prepro-72kDa matrix metalloproteinase; Progelatinase A; TBE1; TBE-1; wu:fa99h12; wu:fk89d01
Gene Symbols MMP2
Host Species Mouse
Immunogen N-terminal peptide APSPIIKFPGD-VAPKTDK of procollagenase IV
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17390, 4313, 81686
Target Species Human, Mouse, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.