Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMP20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102883
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102883 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102883 Supplier Invitrogen™ Supplier No. PA5102883
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MMP20.

MMP20 degrades amelogenin, the major protein component of the enamel matrix, as well as two of the macromolecules characterizing the cartilage extracellular matrix: aggrecan and the cartilage oligomeric matrix protein (COMP). MMP20 may play a central role in tooth enamel formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP20
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MMP20
Gene Accession No. O60882
Gene Alias AI2A2; Enamel metalloproteinase; enamel metalloproteinase precursor; enamelysin; matrix metallo protease; matrix metallopeptidase 20; matrix metallopeptidase 20 (enamelysin); matrix metalloproteinase; matrix metalloproteinase 20; matrix metalloproteinase 20 (enamelysin); matrix metalloproteinase-20; MMP; MMP20; MMP-20; MMPs
Gene Symbols MMP20
Host Species Rabbit
Immunogen A synthesized peptide derived from human MMP20(Accession O60882), corresponding to amino acid residues Q267-D317.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9313
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.