Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | MMP20 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
MMP20 Polyclonal specifically detects MMP20 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MMP20 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
9313 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
AI2A2, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.35, Enamel metalloproteinase, Enamelysin, matrix metallopeptidase 20, matrix metalloproteinase 20 (enamelysin), matrix metalloproteinase-20, MMP-20 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 108-310 of human MMP20 (NP_004762.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title