Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP20 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986803
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6803 Supplier Bioss Supplier No. BS5788R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MMP20 degrades amelogenin, the major protein component of the enamel matrix, as well as two of the macromolecules characterizing the cartilage extracellular matrix: aggrecan and the cartilage oligomeric matrix protein (COMP). MMP20 may play a central role in tooth enamel formation.

Specifications

Antigen MMP20
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MMP20
Gene Accession No. O60882
Gene Alias AI2A2; BOS_14256; Enamel metalloproteinase; Enamelysin; LOC100355428; matrix metallo protease; matrix metallopeptidase 20; matrix metallopeptidase 20 (enamelysin); matrix metallopeptidase 25; matrix metalloproteinase; matrix metalloproteinase 20; matrix metalloproteinase 20 (enamelysin); Matrix metalloproteinase-20; MMP; MMP20; MMP-20; MMP25; MMPs
Gene Symbols MMP20
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MMP20.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9313
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.