Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MMP23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116366
Description
MMP23 Polyclonal Antibody for Western Blot
MMP23B (matrix metallopeptidase 23B) is also named as MMP21, MMP22. It is a regulator in matrix remodeling and may be playing a role in enchondral bone formation and in regulation of cranial suture closure. It has 3 isoforms produced by alternative splicing with the molecular weight of 44 kDa, 48 kDa and 46 kDa, respectively. It also can be N-glycosylated.
Specifications
MMP23 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
O75900 | |
MMP23B | |
KLH conjugated synthetic peptide between 328-362 amino acids from human MMP23A. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MMP23B | |
Femalysin; matrix metallo protease; matrix metallopeptidase 23A (pseudogene); matrix metallopeptidase 23B; matrix metalloproteinase 22; matrix metalloproteinase 23B; matrix metalloproteinase in the female reproductive tract; matrix metalloproteinase-21; Matrix metalloproteinase-22; matrix metalloproteinase-23; Matrix metalloproteinase-23, soluble form; MIFR; MIFR-1; MMP; MMP 23; MMP21; MMP-21; MMP22; MMP-22; MMP23; MMP-23; MMP23A; MMP23B; MMPs; RP11-345P4.2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
8510, 8511 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction