Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MMP24 Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $511.50
Specifications
Antigen | MMP24 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119273
|
Invitrogen™
PA5119273 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121877
|
Invitrogen™
PA5121877 |
20 μL |
Each for $178.40
|
|
|||||
Description
MMP24 Polyclonal Antibody for Western Blot, IHC (P)
All cells within tissues are surrounded by an extracellular matrix (ECM) giving the tissues shape and structure. The ECM is constantly being remodeled and constant communication is maintained between cells through this matrix. Secreted proteins, termed matrix metalloproteinases (MMPs), are involved in the modulation of cell-matrix interactions. MMPs are Zn (2+)-binding endopeptidases that degrade various components of the ECM. MMPs are enzymes implicated in normal and pathologic tissue remodeling processes, wound healing, angiogenesis, and tumor invasion. These enzymes are very potent when active, and are associated with extracellular space inhibitors called TIMPs (tissue inhibitors of matrix metalloproteinases). TIMPs have been shown to block tumor cell invasion suggesting that they act as metastasis suppressor genes.Specifications
MMP24 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Rat | |
MMP24 | |
AU040325; matrix metallo protease; matrix metallopeptidase 24; matrix metallopeptidase 24 (membrane-inserted); matrix metalloproteinase 24; matrix metalloproteinase 24 (membrane-inserted); matrix metalloproteinase-21; Matrix metalloproteinase-24; membrane type 5-MMP; membrane-type 5 matrix metalloproteinase; membrane-type matrix metalloproteinase 5; Membrane-type-5 matrix metalloproteinase; MMP; Mmp21; MMP-21; Mmp24; MMP-24; MMP25; MMPs; MT5MMP; Mt5-mmp; MT-MMP 5; MTMMP5; MT-MMP5; Processed matrix metalloproteinase-24; RP4-614O4.10 | |
MMP24 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q99PW6, Q9Y5R2 | |
10893, 83513 | |
Recombinant protein Matrix Metalloproteinase 24. The antigen corresponds to amino acid range 429-642 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title