Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMP24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100202
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100202 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100202 Supplier Invitrogen™ Supplier No. PA5100202
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MMP24.

All cells within tissues are surrounded by an extracellular matrix (ECM) giving the tissues shape and structure. The ECM is constantly being remodeled and constant communication is maintained between cells through this matrix. Secreted proteins, termed matrix metalloproteinases (MMPs), are involved in the modulation of cell-matrix interactions. MMPs are Zn (2+)-binding endopeptidases that degrade various components of the ECM. MMPs are enzymes implicated in normal and pathologic tissue remodeling processes, wound healing, angiogenesis, and tumor invasion. These enzymes are very potent when active, and are associated with extracellular space inhibitors called TIMPs (tissue inhibitors of matrix metalloproteinases). TIMPs have been shown to block tumor cell invasion suggesting that they act as metastasis suppressor genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP24
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MMP24
Gene Accession No. Q99PW6, Q9R0S2, Q9Y5R2
Gene Alias AU040325; matrix metallo protease; matrix metallopeptidase 24; matrix metallopeptidase 24 (membrane-inserted); matrix metalloproteinase 24; matrix metalloproteinase 24 (membrane-inserted); matrix metalloproteinase-21; Matrix metalloproteinase-24; membrane type 5-MMP; membrane-type 5 matrix metalloproteinase; membrane-type matrix metalloproteinase 5; Membrane-type-5 matrix metalloproteinase; MMP; Mmp21; MMP-21; Mmp24; MMP-24; MMP25; MMPs; MT5MMP; Mt5-mmp; MT-MMP 5; MTMMP5; MT-MMP5; Processed matrix metalloproteinase-24; RP4-614O4.10
Gene Symbols MMP24
Host Species Rabbit
Immunogen A synthesized peptide derived from human MMP24(Accession Q9Y5R2), corresponding to amino acid residues F442-K492.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10893, 17391, 83513
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.