Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MMP25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109967
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109967 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109967 Supplier Invitrogen™ Supplier No. PA5109967
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPRPARLHRF WEGLPAQVRV VQAAYARHRD GRILLFSGPQ FWVFQDRQLE GGARPLTELG LPPGEEVDAV FSWPQNGKTY LVRGRQYWRY DEAAARPDPG YPRDLSLWEG APPSPDDVTV SNAGDTYFFK GAHYWRFPKN SIKTEPDAPQ PMGPNWLDCP APSSGPRAPR PPKATPVSET CDCQCELNQA A.

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. However, the protein encoded by this gene is a member of the membrane-type MMP (MT-MMP) subfamily, attached to the plasma membrane via a glycosylphosphatidyl inositol anchor. In response to bacterial infection or inflammation, the encoded protein is thought to inactivate alpha-1 proteinase inhibitor, a major tissue protectant against proteolytic enzymes released by activated neutrophils, facilitating the transendothelial migration of neutrophils to inflammatory sites. The encoded protein may also play a role in tumor invasion and metastasis through activation of MMP2. The gene has previously been referred to as MMP20 but has been renamed MMP25.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMP25
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Mmp25
Gene Accession No. Q3U435, Q9NPA2
Gene Alias F730048C11Rik; Leukolysin; matrix metallo protease; matrix metallopeptidase 25; matrix metallopeptidase-like 1; matrix metalloproteinase 20; matrix metalloproteinase 25; matrix metalloproteinase-25; matrix metalloproteinase-like 1; membrane-type 6 matrix metalloproteinase; membrane-type matrix metalloproteinase 6; Membrane-type-6 matrix metalloproteinase; MMP; MMP 25; MMP20; MMP20A; Mmp25; MMP-25; MMPL1; MMPs; MT MMP 6; MT6 MMP; MT6MMP; MT6-MMP; MT-MMP 6; MTMMP6; MT-MMP6
Gene Symbols Mmp25
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-540 of human MMP25 (NP_0719131).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240047, 302963, 64386
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.