Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP26 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300124MFV610
Description
MMP26 Polyclonal antibody specifically detects MMP26 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
MMP26 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
EC 3.4.24.-, Endometase, matrilysin 2, matrilysin-2, matrix metallopeptidase 26, matrix metalloproteinase 26, matrix metalloproteinase-26, MGC126590, MGC126592, MMP-26 | |
Produced in rabbits immunized with purified, recombinant Human MMP26 (Accession#: NP_068573.2; Thr90-Pro261) | |
0.1 mL | |
Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
56547 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction